Gene Transcripts
- Gene Symbol
- Hist1h2ae
- Entrez Gene ID
- 319166
- Gene Name
- histone cluster 1, H2ae
- Gene Aliases
- 1190022L06
- Chromosome Location
- Chr. 13: 23570663 - 23571220 on Build GRCm38
- UniGene ID
- Mm.261665
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK028026.1 |
- |
1 |
599 |
|
View Full Product Details