Gene Transcripts

Gene Symbol
Cenpm
Entrez Gene ID
66570
Gene Name
centromere protein M
Gene Aliases
2610019I03Rik, AI853711, Pane1
Chromosome Location
Chr. 15: 82233762 - 82244767 on Build GRCm38
UniGene ID
Mm.23596
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080158.1 NP_001073627.1 5 850
NM_025639.4 NP_079915.1 6 851
NM_178269.3 NP_840000.2 5 759
GenBank mRNA AK010885.1 - 6 843
AK011191.1 - 6 828
AK011227.1 - 6 835
AK011464.1 - 6 852
AK011755.1 - 6 843
AK011793.1 - 6 838
AK017679.1 - 6 844
AK168203.1 - 5 740
BC009160.1 - 6 833 2655787
BY710734.1 - 6 851

View Full Product Details