Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_201643.2 | NP_964005.2 | 1 | 118 | |
GenBank mRNA | AB094481.1 | - | 1 | 97 | |
AY227196.1 | - | 1 | 118 | ||
BC139093.1 | - | 1 | 98 | 8862115 | |
BC139094.1 | - | 1 | 98 | 8862116 |