Gene Transcripts

Gene Symbol
KIAA1524
Entrez Gene ID
57650
Gene Name
KIAA1524
Gene Aliases
CIP2A, p90
Chromosome Location
Chr. 3: 108545752 - 108589644 on Build GRCh38
UniGene ID
Hs.591308
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020890.2 NP_065941.2 8 1150
XM_006713716.3 XP_006713779.1 8 934
XM_006713717.2 XP_006713780.1 9 1046
XM_011513056.1 XP_011511358.1 8 894
XM_011513057.2 XP_011511359.1 9 1046
GenBank mRNA AB040957.1 - 8 877
AF334474.1 - 8 949
AK056933.1 - 8 959
AK308315.1 - 8 896
AK310446.1 - 8 1154
AL713689.1 - 8 945
BC063433.1 - 8 941
BC082961.1 - 8 903
BC130564.1 - 8 917 40146595
BC136371.1 - 8 917 9020358

View Full Product Details