Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020962.2 | NP_066013.1 | 3 | 752 | |
XM_011521845.2 | XP_011520147.1 | 3 | 932 | ||
XM_011521846.2 | XP_011520148.1 | 3 | 556 | ||
XM_011521847.1 | XP_011520149.1 | 3 | 616 | ||
XM_017022448.1 | XP_016877937.1 | 3 | 934 | ||
XM_017022449.1 | XP_016877938.1 | 3 | 940 | ||
GenBank mRNA | AB052622.1 | - | 3 | 752 |