Gene Transcripts

Gene Symbol
CDKL4
Entrez Gene ID
344387
Gene Name
cyclin dependent kinase like 4
Gene Aliases
Chromosome Location
Chr. 2: 39174421 - 39246815 on Build GRCh38
UniGene ID
Hs.403201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009565.1 NP_001009565.1 1 82
XM_011532815.2 XP_011531117.1 2 1498
XM_011532816.2 XP_011531118.1 2 1498
XM_011532817.2 XP_011531119.1 2 1498
XM_011532819.2 XP_011531121.1 2 1498
XM_017003981.1 XP_016859470.1 2 850
XM_017003982.1 XP_016859471.1 2 326
XM_017003983.1 XP_016859472.1 2 1498
XM_017003984.1 XP_016859473.1 2 1498
XM_017003985.1 XP_016859474.1 2 1498
GenBank mRNA AY845084.1 - 1 82
AY847283.1 - 1 82
BC144460.1 - 1 146

View Full Product Details