Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003041.3 | NP_003032.1 | 1 | 109 | |
NR_130783.1 | 1 | 109 | |||
XM_006721072.3 | XP_006721135.2 | 1 | 692 | ||
GenBank mRNA | BC131542.1 | - | 1 | 109 | |
M95549.1 | - | 1 | 110 |