Gene Transcripts

Gene Symbol
GPR31
Entrez Gene ID
2853
Gene Name
G protein-coupled receptor 31
Gene Aliases
12-HETER, HETER, HETER1
Chromosome Location
Chr. 6: 167156872 - 167157831 on Build GRCh38
UniGene ID
Hs.248124
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005299.2 NP_005290.2 1 586
GenBank mRNA BC095537.1 - 1 583
CD246167.1 - 1 594

View Full Product Details