Gene Transcripts

Gene Symbol
KLHL8
Entrez Gene ID
57563
Gene Name
kelch like family member 8
Gene Aliases
Chromosome Location
Chr. 4: 87160103 - 87220608 on Build GRCh38
UniGene ID
Hs.106601
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001292003.1 NP_001278932.1 2 424
NM_001292006.1 NP_001278935.1 2 381
NM_001292007.1 NP_001278936.1 2 381
NM_020803.4 NP_065854.3 2 381
NR_120447.1 2 381
GenBank mRNA AB037799.2 - 1 64
AK128827.1 - 2 424
BC041384.2 - 2 331 5275946
BC041901.1 - 2 237
BX538173.1 - 2 298
BX640727.1 - 2 291
BX640744.1 - 2 310
BX647454.1 - 2 316
CN290430.1 - 2 396
CR749265.1 - 2 299

View Full Product Details