Gene Transcripts

Gene Symbol
ESCO1
Entrez Gene ID
114799
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 1
Gene Aliases
A930014I12Rik, CTF, ECO1, EFO1, ESO1
Chromosome Location
Chr. 18: 21529281 - 21601015 on Build GRCh38
UniGene ID
Hs.464733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052911.2 NP_443143.2 4 970
XM_011525798.1 XP_011524100.1 4 1100
XM_011525799.2 XP_011524101.1 4 1233
GenBank mRNA AB067498.1 - 4 906
AK096746.1 - 4 973
AL832041.1 - 4 1123
BC089426.1 - 1 390 3090307

View Full Product Details