Gene Transcripts

Gene Symbol
RAB15
Entrez Gene ID
376267
Gene Name
RAB15, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 14: 64945814 - 64972197 on Build GRCh38
UniGene ID
Hs.512492
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308154.1 NP_001295083.1 7 2221
NM_198686.2 NP_941959.1 7 2352
XM_017021277.1 XP_016876766.1 8 2481
GenBank mRNA BC040679.2 - 7 2274
BX248046.1 - 6 2148

View Full Product Details