Gene Transcripts
- Gene Symbol
- PI4KAP2
- Entrez Gene ID
- 375133
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 2
- Gene Aliases
- Chromosome Location
- Chr. 22: 21472998 - 21517491 on Build GRCh38
- UniGene ID
- Hs.448225
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003700.1 |
|
17 |
2119 |
|
GenBank mRNA |
AK298744.1 |
- |
17 |
2261 |
|
BC020225.1 |
- |
17 |
2118 |
|
- Gene Symbol
- PI4KAP1
- Entrez Gene ID
- 728233
- Gene Name
- phosphatidylinositol 4-kinase alpha pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 22: 18533864 - 18548828 on Build GRCh38
- UniGene ID
- Hs.592456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK002141.1 |
- |
3 |
2272 |
|
AK024034.1 |
- |
4 |
1645 |
|
View Full Product Details