Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271028.1 | NP_001257957.1 | 2 | 467 | |
NM_021153.3 | NP_066976.1 | 2 | 467 | ||
NR_073130.1 | 2 | 467 | |||
XM_011525931.2 | XP_011524233.1 | 2 | 570 | ||
XM_011525932.1 | XP_011524234.1 | 1 | 245 | ||
XM_017025711.1 | XP_016881200.1 | 2 | 492 | ||
GenBank mRNA | AJ007607.1 | - | 2 | 308 | |
AK304000.1 | - | 2 | 417 | ||
AK315257.1 | - | 2 | 469 | ||
AY358654.1 | - | 1 | 206 | ||
BC015877.1 | - | 2 | 359 | 4716394 |