Gene Transcripts
- Gene Symbol
- EXOC4
- Entrez Gene ID
- 60412
- Gene Name
- exocyst complex component 4
- Gene Aliases
- SEC8, SEC8L1, Sec8p
- Chromosome Location
- Chr. 7: 133253067 - 134067137 on Build GRCh38
- UniGene ID
- Hs.321273
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001037126.1 |
NP_001032203.1
|
1, 2 |
129 |
|
NM_021807.3 |
NP_068579.3
|
1, 2 |
129 |
|
XM_005250523.4 |
XP_005250580.1
|
1, 2 |
113 |
|
XM_017012495.1 |
XP_016867984.1
|
1, 2 |
113 |
|
GenBank mRNA |
AB051486.1 |
- |
1, 2 |
71 |
|
AF380839.1 |
- |
1, 2 |
126 |
|
AK027688.1 |
- |
1, 2 |
123 |
|
AL831989.2 |
- |
1, 2 |
124 |
|
BC026174.1 |
- |
1, 2 |
105 |
|
BC067263.1 |
- |
1, 2 |
106 |
6183004 |
BE244117.1 |
- |
1, 2 |
152 |
|
BE246205.1 |
- |
1, 2 |
152 |
|
View Full Product Details