Gene Transcripts

Gene Symbol
DUS1L
Entrez Gene ID
64118
Gene Name
dihydrouridine synthase 1 like
Gene Aliases
DUS1, PP3111
Chromosome Location
Chr. 17: 82057872 - 82065821 on Build GRCh38
UniGene ID
Hs.514599
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022156.4 NP_071439.3 8 989
XM_005256393.2 XP_005256450.1 8 983
XM_005256394.2 XP_005256451.1 8 983
XM_006722288.2 XP_006722351.1 7 932
XM_006722289.2 XP_006722352.1 7 932
GenBank mRNA AF202637.1 - 2 127
AK226154.1 - 3 1141
AK298330.1 - 6 560
BC003659.2 - 4 379
BC017081.1 - 7 570
BC062566.1 - 7 1189 5171876
BC064918.1 - 8 807 6092677

View Full Product Details