Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022167.3 | NP_071450.2 | 2 | 303 | |
NR_110010.1 | 2 | 303 | |||
XM_005257572.4 | XP_005257629.1 | 2 | 617 | ||
GenBank mRNA | AJ277442.1 | - | 2 | 303 | |
AK299999.1 | - | 2 | 200 | ||
AY358090.1 | - | 2 | 199 | ||
BC052262.1 | - | 2 | 223 | 6380013 |