Gene Transcripts

Gene Symbol
FAM173A
Entrez Gene ID
65990
Gene Name
family with sequence similarity 173 member A
Gene Aliases
C16orf24
Chromosome Location
Chr. 16: 720581 - 722590 on Build GRCh38
UniGene ID
Hs.166244
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023933.2 NP_076422.1 3, 4 537
GenBank mRNA BC001181.1 - 3, 4 521
BC002624.2 - 3, 4 489 3606040

View Full Product Details