Gene Transcripts
- Gene Symbol
- CENPM
- Entrez Gene ID
- 79019
- Gene Name
- centromere protein M
- Gene Aliases
- C22orf18, CENP-M, PANE1
- Chromosome Location
- Chr. 22: 41927747 - 41947164 on Build GRCh38
- UniGene ID
- Hs.208912
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002876.2 |
NP_001002876.1
|
2, 3 |
231 |
|
NM_001304370.1 |
NP_001291299.1
|
1, 2 |
344 |
|
NM_001304371.1 |
NP_001291300.1
|
2, 3 |
231 |
|
NM_001304372.1 |
NP_001291301.1
|
1, 2 |
344 |
|
NM_001304373.1 |
NP_001291302.1
|
1, 2 |
344 |
|
NM_024053.4 |
NP_076958.1
|
2, 3 |
231 |
|
XM_011530368.2 |
XP_011528670.1
|
2, 3 |
218 |
|
GenBank mRNA |
AK123479.1 |
- |
2, 3 |
220 |
|
AL160131.1 |
- |
2, 3 |
179 |
|
BC000705.2 |
- |
2, 3 |
178 |
3349507 |
BC007495.2 |
- |
2, 3 |
191 |
2966541 |
BE789455.1 |
- |
1, 2 |
274 |
|
BE797487.1 |
- |
2, 3 |
185 |
|
BM045658.1 |
- |
1, 2 |
319 |
|
BQ652721.1 |
- |
1, 2 |
181 |
|
BQ922403.1 |
- |
2, 3 |
164 |
|
BU556693.1 |
- |
2, 3 |
231 |
|
BX332140.2 |
- |
1, 2 |
282 |
|
BX354019.2 |
- |
1, 2 |
278 |
|
BX417558.2 |
- |
1, 2 |
283 |
|
CR456399.1 |
- |
2, 3 |
204 |
|
CR457392.1 |
- |
2, 3 |
143 |
|
HY095078.1 |
- |
1, 2 |
345 |
|
View Full Product Details