Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136201.1 | NP_001129673.1 | 5, 6 | 716 | |
NM_001136202.1 | NP_001129674.1 | 4, 5 | 506 | ||
NM_024710.2 | NP_078986.1 | 5, 6 | 764 | ||
GenBank mRNA | AK027122.1 | - | 5, 6 | 727 | |
AK131324.1 | - | 4, 5 | 1167 | ||
BC017344.2 | - | 5, 6 | 603 | 4893552 | |
CR457377.1 | - | 4, 5 | 590 |