Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318007.1 | NP_001304936.1 | 3, 4 | 399 | |
NM_024792.2 | NP_079068.1 | 4, 5 | 601 | ||
GenBank mRNA | AF477201.1 | - | 4, 5 | 589 | |
AK025935.1 | - | 4, 5 | 540 | ||
AK125146.1 | - | 3, 4 | 806 | ||
AK292065.1 | - | 4, 5 | 540 | ||
BC009729.1 | - | 1, 2 | 94 | ||
BX389422.1 | - | 2, 3 | 146 |