Gene Transcripts
- Gene Symbol
- TMC7
- Entrez Gene ID
- 79905
- Gene Name
- transmembrane channel like 7
- Gene Aliases
- Chromosome Location
- Chr. 16: 18983934 - 19063942 on Build GRCh38
- UniGene ID
- Hs.187377
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001160364.2 |
NP_001153836.1
|
8, 9 |
1415 |
|
NM_001300732.1 |
NP_001287661.1
|
8, 9 |
1317 |
|
NM_001324263.1 |
NP_001311192.1
|
8, 9 |
1304 |
|
NM_001324265.1 |
NP_001311194.1
|
8, 9 |
1317 |
|
NM_001324268.1 |
NP_001311197.1
|
7, 8 |
1169 |
|
NM_024847.4 |
NP_079123.3
|
8, 9 |
1317 |
|
NR_136733.1 |
|
8, 9 |
1304 |
|
GenBank mRNA |
AK024893.1 |
- |
4, 5 |
526 |
|
AK094664.1 |
- |
8, 9 |
1317 |
|
AK293874.1 |
- |
8, 9 |
1211 |
|
AY236498.1 |
- |
8, 9 |
1251 |
|
AY263165.1 |
- |
8, 9 |
1246 |
|
BC036205.1 |
- |
8, 9 |
1246 |
5261157 |
BC047719.1 |
- |
8, 9 |
1248 |
5743899 |
CR933669.1 |
- |
8, 9 |
1415 |
|
View Full Product Details