Gene Transcripts

Gene Symbol
POF1B
Entrez Gene ID
79983
Gene Name
premature ovarian failure, 1B
Gene Aliases
POF, POF2B
Chromosome Location
Chr. X: 85277389 - 85379743 on Build GRCh38
UniGene ID
Hs.267038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001307940.1 NP_001294869.1 15 1782
NM_024921.3 NP_079197.3 15 1782
XM_005262203.3 XP_005262260.1 15 1695
GenBank mRNA AF309774.1 - 15 1782
AK025039.1 - 11 1375
AK025080.1 - 7 747
AK026445.1 - 3 275
AK225193.1 - 7 747
AK290360.1 - 15 1731
AK314295.1 - 15 1701
BC017500.1 - 15 1756 4905554

View Full Product Details