Gene Transcripts

Gene Symbol
FOXN3
Entrez Gene ID
1112
Gene Name
forkhead box N3
Gene Aliases
C14orf116, CHES1, PRO1635
Chromosome Location
Chr. 14: 89156172 - 89619150 on Build GRCh38
UniGene ID
Hs.434286
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001085471.1 NP_001078940.1 2 559
NM_005197.3 NP_005188.2 2 676
GenBank mRNA AK225967.1 - 4 410
AK308053.1 - 2 558
BC007506.2 - 2 570 3028787
BP213901.1 - 4 429
U68723.1 - 2 554

View Full Product Details