Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308164.1 | NP_001295093.1 | 17 | 2218 | |
NM_024996.5 | NP_079272.4 | 16 | 2161 | ||
XM_006713795.1 | XP_006713858.1 | 15 | 2075 | ||
GenBank mRNA | AF309777.1 | - | 16 | 2161 | |
AF367998.1 | - | 16 | 2161 | ||
AK092293.1 | - | 15 | 1767 | ||
AK315031.1 | - | 16 | 2100 | ||
BC049210.1 | - | 16 | 2125 | 5574223 | |
KU178816.1 | - | 16 | 2054 | ||
KU178817.1 | - | 15 | 1933 |