Gene Transcripts

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
Chromosome Location
Chr. 20: 3667973 - 3682131 on Build GRCh38
UniGene ID
Hs.173716
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282447.2 NP_001269376.1 6 1125
NM_025220.4 NP_079496.1 6 1125
NM_153202.3 NP_694882.1 6 1125
XM_005260843.1 XP_005260900.1 6 582
XM_006723639.1 XP_006723702.1 6 582
XM_006723640.1 XP_006723703.1 6 582
XM_006723644.2 XP_006723707.1 6 582
XM_011529366.1 XP_011527668.1 6 481
XM_011529367.1 XP_011527669.1 6 442
XM_011529368.1 XP_011527670.1 6 582
XM_011529370.1 XP_011527672.1 6 582
XM_011529371.1 XP_011527673.1 6 582
XM_011529373.1 XP_011527675.1 6 479
XM_017028080.1 XP_016883569.1 6 582
XM_017028081.1 XP_016883570.1 6 503
XM_017028082.1 XP_016883571.1 6 582
XM_017028083.1 XP_016883572.1 6 582
GenBank mRNA AB055891.1 - 6 526
AF466287.1 - 6 444
AK123015.1 - 3 605
AK126487.1 - 4 1381
AK126498.1 - 4 287
AK226148.1 - 5 829
AK300429.1 - 6 493
AK304037.1 - 6 479
AY358314.1 - 6 552
BC125112.1 - 4 742
BC125113.1 - 6 486 40118831
DB225158.1 - 6 503

View Full Product Details