Gene Transcripts

Gene Symbol
TUBB1
Entrez Gene ID
81027
Gene Name
tubulin beta 1 class VI
Gene Aliases
Chromosome Location
Chr. 20: 59016026 - 59026654 on Build GRCh38
UniGene ID
Hs.303023
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030773.3 NP_110400.1 4 1284
XM_017028085.1 XP_016883574.1 4 1478
GenBank mRNA AJ292757.1 - 4 1284
BC033679.1 - 4 1117 5180793

View Full Product Details