Gene Transcripts
- Gene Symbol
- C1QTNF1
- Entrez Gene ID
- 114897
- Gene Name
- C1q and tumor necrosis factor related protein 1
- Gene Aliases
- CTRP1, GIP, ZSIG37
- Chromosome Location
- Chr. 17: 79022934 - 79049788 on Build GRCh38
- UniGene ID
- Hs.201398
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030968.3 |
NP_112230.1
|
2, 3 |
596 |
|
NM_153372.1 |
NP_699203.1
|
2, 3 |
332 |
|
NM_198593.3 |
NP_940995.1
|
2, 3 |
412 |
|
NM_198594.2 |
NP_940996.1
|
1, 2 |
427 |
|
NR_049769.1 |
|
2, 3 |
450 |
|
XM_006721663.1 |
XP_006721726.1
|
1, 2 |
457 |
|
XM_006721664.1 |
XP_006721727.1
|
4, 5 |
1044 |
|
XM_006721666.1 |
XP_006721729.1
|
3, 4 |
660 |
|
XM_017024126.1 |
XP_016879615.1
|
1, 2 |
701 |
|
GenBank mRNA |
AF232905.1 |
- |
1, 2 |
539 |
|
AF329840.1 |
- |
2, 3 |
333 |
|
AJ272138.1 |
- |
1, 2 |
539 |
|
AK055541.1 |
- |
1, 2 |
242 |
|
AK172767.1 |
- |
2, 3 |
433 |
|
AK292034.1 |
- |
2, 3 |
450 |
|
AY358424.1 |
- |
2, 3 |
562 |
|
BC021553.1 |
- |
2, 3 |
218 |
4622952 |
DA877694.1 |
- |
2, 3 |
332 |
|
DB126803.1 |
- |
2, 3 |
412 |
|
View Full Product Details