Gene Transcripts

Gene Symbol
KAZALD1
Entrez Gene ID
81621
Gene Name
Kazal type serine peptidase inhibitor domain 1
Gene Aliases
BONO1, FKSG28, FKSG40, IGFBP-rP10
Chromosome Location
Chr. 10: 101061141 - 101068141 on Build GRCh38
UniGene ID
Hs.534859
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319303.1 NP_001306232.1 5 1870
NM_030929.4 NP_112191.2 5 1815
NR_135067.1 4 1253
XM_017016714.1 XP_016872203.1 4 894
XM_017016715.1 XP_016872204.1 4 894
GenBank mRNA AK172862.1 - 5 1066
AK172864.1 - 5 1869
AK225949.1 - 5 1067
AY359087.1 - 5 1071
BC007758.2 - 5 1216 4122366
BG034107.1 - 1 79
BI766345.1 - 4 526
CX871488.1 - 4 544

View Full Product Details