Gene Transcripts

Gene Symbol
RAB6C
Entrez Gene ID
84084
Gene Name
RAB6C, member RAS oncogene family
Gene Aliases
WTH3
Chromosome Location
Chr. 2: 129979662 - 129982738 on Build GRCh38
UniGene ID
Hs.591552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032144.2 NP_115520.2 1 901
GenBank mRNA AF309646.1 - 1 494
AK055504.1 - 2 288
AL136727.1 - 1 902
BC120999.1 - 1 739 40116526
CR533469.1 - 1 447

Gene Symbol
WTH3DI
Entrez Gene ID
150786
Gene Name
RAB6C-like
Gene Aliases
Chromosome Location
Chr. 2: 131360492 - 131364158 on Build GRCh38
UniGene ID
Hs.645670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077637.1 NP_001071105.1 1 885
GenBank mRNA AB232598.1 - 1 447

View Full Product Details