Gene Transcripts
- Gene Symbol
- FAM213A
- Entrez Gene ID
- 84293
- Gene Name
- family with sequence similarity 213 member A
- Gene Aliases
- C10orf58, PAMM
- Chromosome Location
- Chr. 10: 80407829 - 80437115 on Build GRCh38
- UniGene ID
- Hs.500333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243778.1 |
NP_001230707.1
|
2, 3 |
245 |
|
NM_001243779.1 |
NP_001230708.1
|
2, 3 |
315 |
|
NM_001243780.1 |
NP_001230709.1
|
2, 3 |
811 |
|
NM_001243781.1 |
NP_001230710.1
|
2, 3 |
328 |
|
NM_001243782.1 |
NP_001230711.1
|
2, 3 |
228 |
|
NM_032333.4 |
NP_115709.3
|
2, 3 |
345 |
|
XM_011540265.2 |
XP_011538567.1
|
2, 3 |
648 |
|
XM_011540266.2 |
XP_011538568.1
|
2, 3 |
1051 |
|
XM_017016775.1 |
XP_016872264.1
|
2, 3 |
332 |
|
GenBank mRNA |
AK074643.1 |
- |
2, 3 |
290 |
|
AK075447.1 |
- |
2, 3 |
320 |
|
AK225387.1 |
- |
2, 3 |
219 |
|
AK315440.1 |
- |
2, 3 |
239 |
|
AY359059.1 |
- |
2, 3 |
238 |
|
BC005871.2 |
- |
2, 3 |
207 |
3010078 |
BE742158.1 |
- |
2, 3 |
315 |
|
CN396006.1 |
- |
2, 3 |
197 |
|
DA136257.1 |
- |
2, 3 |
229 |
|
DB494833.1 |
- |
2, 3 |
345 |
|
View Full Product Details