Gene Transcripts
- Gene Symbol
- GLYR1
- Entrez Gene ID
- 84656
- Gene Name
- glyoxylate reductase 1 homolog
- Gene Aliases
- BM045, HIBDL, N-PAC, NP60
- Chromosome Location
- Chr. 16: 4803203 - 4847388 on Build GRCh38
- UniGene ID
- Hs.387255
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308096.1 |
NP_001295025.1
|
2, 3 |
115 |
|
NM_001324096.1 |
NP_001311025.1
|
2, 3 |
201 |
|
NM_001324097.1 |
NP_001311026.1
|
2, 3 |
201 |
|
NM_001324098.1 |
NP_001311027.1
|
2, 3 |
201 |
|
NM_032569.3 |
NP_115958.2
|
2, 3 |
115 |
|
NR_136695.1 |
|
2, 3 |
201 |
|
NR_136696.1 |
|
2, 3 |
201 |
|
NR_136697.1 |
|
2, 3 |
201 |
|
NR_136698.1 |
|
2, 3 |
201 |
|
NR_136699.1 |
|
2, 3 |
201 |
|
NR_136700.1 |
|
2, 3 |
201 |
|
XM_005255639.4 |
XP_005255696.2
|
2, 3 |
159 |
|
XM_011522716.2 |
XP_011521018.1
|
2, 3 |
243 |
|
XM_011522717.1 |
XP_011521019.1
|
2, 3 |
101 |
|
XM_017023786.1 |
XP_016879275.1
|
2, 3 |
196 |
|
XM_017023787.1 |
XP_016879276.1
|
2, 3 |
196 |
|
XM_017023788.1 |
XP_016879277.1
|
2, 3 |
101 |
|
XM_017023789.1 |
XP_016879278.1
|
2, 3 |
101 |
|
GenBank mRNA |
AF244907.1 |
- |
2, 3 |
96 |
|
AF326966.1 |
- |
2, 3 |
78 |
|
AK296842.1 |
- |
2, 3 |
92 |
|
AK307834.1 |
- |
2, 3 |
91 |
|
AY352585.1 |
- |
2, 3 |
78 |
|
BC003693.1 |
- |
1, 2 |
40 |
|
BC032855.1 |
- |
2, 3 |
106 |
5265561 |
BC047223.1 |
- |
2, 3 |
95 |
5263991 |
BC064940.1 |
- |
2, 3 |
100 |
5787345 |
DB492328.1 |
- |
2, 3 |
120 |
|
HY130375.1 |
- |
2, 3 |
202 |
|
View Full Product Details