Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278270.1 | NP_001265199.1 | 3 | 460 | |
NM_006354.3 | NP_006345.1 | 3 | 1066 | ||
NM_133480.2 | NP_597814.1 | 3 | 1066 | ||
GenBank mRNA | AF069733.1 | - | 2 | 243 | |
AK000228.1 | - | 3 | 470 | ||
AK292264.1 | - | 3 | 460 | ||
AL117487.1 | - | 3 | 729 | ||
BC009240.2 | - | 3 | 791 | 3009860 | |
BC013433.1 | - | 3 | 729 | 3897746 | |
CR533543.1 | - | 2 | 243 | ||
DB032623.1 | - | 4 | 328 |