Gene Transcripts

Gene Symbol
KLF12
Entrez Gene ID
11278
Gene Name
Kruppel like factor 12
Gene Aliases
AP-2rep, AP2REP, HSPC122
Chromosome Location
Chr. 13: 73686012 - 74168295 on Build GRCh38
UniGene ID
Hs.373857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007249.4 NP_009180.3 4 699
XM_005266251.3 XP_005266308.1 4 808
XM_011534907.2 XP_011533209.1 3 522
XM_011534908.2 XP_011533210.1 3 522
XM_011534909.2 XP_011533211.1 4 655
XM_011534910.2 XP_011533212.1 3 486
XM_011534911.2 XP_011533213.1 3 564
XM_011534912.2 XP_011533214.1 3 635
XM_017020384.1 XP_016875873.1 3 466
XM_017020385.1 XP_016875874.1 4 739
GenBank mRNA AF113122.1 - 4 673
AF161471.1 - 4 547
AF330041.1 - 3 485
AJ243274.1 - 3 477
AK291397.1 - 4 795
BC019680.1 - 3 504 4942405
DB125574.1 - 4 544
HF546212.1 - 4 670

View Full Product Details