Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001172703.2 | NP_001166174.1 | 2 | 149 | |
NM_001172704.2 | NP_001166175.1 | 2 | 149 | ||
NM_001291641.1 | NP_001278570.1 | 3 | 320 | ||
NM_014001.4 | NP_054720.1 | 2 | 151 | ||
NM_138619.3 | NP_619525.1 | 2 | 151 | ||
XM_011524563.2 | XP_011522865.1 | 2 | 125 | ||
XM_017024385.1 | XP_016879874.1 | 2 | 125 | ||
XM_017024386.1 | XP_016879875.1 | 2 | 113 | ||
XM_017024387.1 | XP_016879876.1 | 3 | 274 | ||
XM_017024388.1 | XP_016879877.1 | 3 | 286 | ||
XM_017024389.1 | XP_016879878.1 | 2 | 119 | ||
XM_017024390.1 | XP_016879879.1 | 2 | 117 | ||
XM_017024391.1 | XP_016879880.1 | 2 | 108 | ||
XM_017024392.1 | XP_016879881.1 | 3 | 276 | ||
GenBank mRNA | AF190864.1 | - | 2 | 85 | |
AF219138.1 | - | 2 | 86 | ||
AF219139.1 | - | 2 | 86 | ||
AK291651.1 | - | 2 | 116 | ||
AK291685.1 | - | 2 | 82 | ||
AK293291.1 | - | 2 | 116 | ||
AK301895.1 | - | 2 | 117 | ||
AK302278.1 | - | 2 | 119 | ||
AK302423.1 | - | 2 | 80 | ||
AK304692.1 | - | 2 | 80 | ||
AL833603.1 | - | 3 | 260 | ||
BC063285.1 | - | 2 | 113 | ||
BC070044.1 | - | 2 | 90 | 4814878 | |
D63876.1 | - | 2 | 85 | ||
DA312660.1 | - | 2 | 151 | ||
HY030361.1 | - | 3 | 319 |