Gene Transcripts

Gene Symbol
IQCD
Entrez Gene ID
115811
Gene Name
IQ motif containing D
Gene Aliases
4933433C09Rik, CFAP84, DRC10
Chromosome Location
Chr. 12: 113195441 - 113221344 on Build GRCh38
UniGene ID
Hs.568276
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138451.2 NP_612460.1 2 838
XM_005253832.4 XP_005253889.1 2 1081
XM_005253833.4 XP_005253890.1 2 1329
XM_011537862.2 XP_011536164.1 2 1961
XM_011537863.2 XP_011536165.1 3 1207
XM_011537864.1 XP_011536166.1 2 838
GenBank mRNA AK127152.1 - 2 838
BC013151.1 - 2 819 4385421

View Full Product Details