Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040272.5 | NP_001035362.3 | 4 | 539 | |
NM_052866.4 | NP_443098.3 | 4 | 539 | ||
XM_011518063.2 | XP_011516365.1 | 6 | 677 | ||
XM_011518064.2 | XP_011516366.1 | 5 | 792 | ||
XM_017015310.1 | XP_016870799.1 | 5 | 7314 | ||
XM_017015311.1 | XP_016870800.1 | 6 | 677 | ||
XM_017015312.1 | XP_016870801.1 | 5 | 793 | ||
XM_017015313.1 | XP_016870802.1 | 5 | 440 | ||
XM_017015314.1 | XP_016870803.1 | 6 | 677 | ||
GenBank mRNA | AF176313.1 | - | 4 | 437 | |
AK290667.1 | - | 4 | 525 | ||
AK291956.1 | - | 4 | 479 | ||
AY358327.1 | - | 4 | 519 | ||
BC030262.1 | - | 4 | 451 | 5201199 | |
CX758410.1 | - | 4 | 438 | ||
DA598045.1 | - | 4 | 514 |