Gene Transcripts

Gene Symbol
LGI4
Entrez Gene ID
163175
Gene Name
leucine rich repeat LGI family member 4
Gene Aliases
LGIL3
Chromosome Location
Chr. 19: 35124513 - 35135274 on Build GRCh38
UniGene ID
Hs.65256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139284.2 NP_644813.1 9 2433
XM_011526595.1 XP_011524897.1 10 1914
XM_017026428.1 XP_016881917.1 9 1762
XM_017026429.1 XP_016881918.1 10 1834
XM_017026430.1 XP_016881919.1 8 1616
GenBank mRNA AJ487519.1 - 9 2358
AJ487959.1 - 9 1866
AL832968.1 - 6 2334
BC029943.1 - 10 1898
BC136694.1 - 9 1908 9020684
BC136697.1 - 9 1908 9020687

View Full Product Details