Gene Transcripts

Gene Symbol
DPP9
Entrez Gene ID
91039
Gene Name
dipeptidyl peptidase 9
Gene Aliases
DP9, DPLP9, DPRP-2, DPRP2
Chromosome Location
Chr. 19: 4675226 - 4723843 on Build GRCh38
UniGene ID
Hs.515081
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139159.4 NP_631898.3 14 1805
XM_005259673.2 XP_005259730.1 15 1914
XM_011528405.1 XP_011526707.1 16 1966
XM_011528406.1 XP_011526708.1 17 2076
XM_011528408.1 XP_011526710.1 14 1751
XM_011528410.1 XP_011526712.1 15 1966
XM_017027453.1 XP_016882942.1 16 2556
XM_017027454.1 XP_016882943.1 17 2666
GenBank mRNA AF452102.2 - 14 1825
AF542510.2 - 14 1784
AK054656.1 - 3 345
AK122645.1 - 8 855
AK122654.1 - 14 1808
AK131100.1 - 11 1374
AK131499.1 - 13 1318
AL834376.1 - 8 836
AY172660.1 - 11 1470
AY374518.1 - 11 1460
BC000970.1 - 2 188
BC037948.2 - 14 1742 4577856
CR627380.1 - 6 580
DQ417928.1 - 12 1547

View Full Product Details