Gene Transcripts

Gene Symbol
CARNMT1
Entrez Gene ID
138199
Gene Name
carnosine N-methyltransferase 1
Gene Aliases
C9orf41, UPF0586
Chromosome Location
Chr. 9: 74980794 - 75028456 on Build GRCh38
UniGene ID
Hs.567688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152420.2 NP_689633.1 1 275
NR_135282.1 1 275
XM_011518223.2 XP_011516525.1 1 301
GenBank mRNA AK299691.1 - 1 118
BC034033.1 - 1 239 4278547
BI560441.1 - 1 280
BX647386.1 - 1 44

View Full Product Details