Gene Transcripts

Gene Symbol
UGT3A1
Entrez Gene ID
133688
Gene Name
UDP glycosyltransferase family 3 member A1
Gene Aliases
Chromosome Location
Chr. 5: 35953089 - 36001055 on Build GRCh38
UniGene ID
Hs.254699
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152404.3 NP_689617.3 7 1754
XM_005248243.4 XP_005248300.1 7 1710
XM_011513957.2 XP_011512259.1 7 1483
XM_011513958.2 XP_011512260.1 6 1840
XM_011513959.2 XP_011512261.1 8 1646
XM_011513961.2 XP_011512263.1 6 1403
GenBank mRNA AK057066.1 - 7 1718
AK091977.1 - 4 737
AK125803.1 - 8 3897
AK290809.1 - 7 1711
AK308046.1 - 7 1660
AK315851.1 - 7 1718
BC068446.1 - 7 1621 5295502

View Full Product Details