Gene Transcripts
- Gene Symbol
- DCLK2
- Entrez Gene ID
- 166614
- Gene Name
- doublecortin like kinase 2
- Gene Aliases
- CL2, CLICK-II, CLICK2, CLIK2, DCAMKL2, DCDC3, DCDC3B, DCK2
- Chromosome Location
- Chr. 4: 150078274 - 150258563 on Build GRCh38
- UniGene ID
- Hs.591683
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040260.3 |
NP_001035350.2
|
1, 2 |
1183 |
|
NM_001040261.4 |
NP_001035351.4
|
1, 2 |
1183 |
|
NR_036614.1 |
|
1, 2 |
1183 |
|
XM_005262785.2 |
XP_005262842.1
|
1, 2 |
1012 |
|
XM_005262786.2 |
XP_005262843.1
|
1, 2 |
1012 |
|
XM_017007832.1 |
XP_016863321.1
|
1, 2 |
1012 |
|
XM_017007833.1 |
XP_016863322.1
|
1, 2 |
1012 |
|
GenBank mRNA |
AB209181.1 |
- |
1, 2 |
1183 |
|
AL834498.1 |
- |
1, 2 |
230 |
|
BC032726.1 |
- |
1, 2 |
476 |
|
View Full Product Details