Gene Transcripts

Gene Symbol
SLC51A
Entrez Gene ID
200931
Gene Name
solute carrier family 51 alpha subunit
Gene Aliases
OSTA, OSTalpha
Chromosome Location
Chr. 3: 196216512 - 196233430 on Build GRCh38
UniGene ID
Hs.630585
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152672.5 NP_689885.4 7, 8 989
GenBank mRNA AK172837.1 - 7, 8 992
AK310276.1 - 4, 5 640
AY194243.1 - 7, 8 810
BC029606.1 - 3, 4 601 5297443

View Full Product Details