Gene Transcripts

Gene Symbol
LACC1
Entrez Gene ID
144811
Gene Name
laccase domain containing 1
Gene Aliases
C13orf31
Chromosome Location
Chr. 13: 43878834 - 43893932 on Build GRCh38
UniGene ID
Hs.210586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128303.1 NP_001121775.1 2 607
NM_153218.2 NP_694950.2 2 443
XM_005266261.3 XP_005266318.1 2 1066
XM_006719766.3 XP_006719829.1 2 224
XM_006719767.3 XP_006719830.1 2 443
XM_011534933.2 XP_011533235.1 3 1261
XM_011534934.2 XP_011533236.1 2 380
XM_011534935.2 XP_011533237.1 2 443
XM_017020394.1 XP_016875883.1 2 500
GenBank mRNA AK096044.1 - 2 443
BC035749.1 - 2 412 5750179
BX503941.1 - 2 334
DR001026.1 - 2 334

View Full Product Details