Gene Transcripts

Gene Symbol
PI16
Entrez Gene ID
221476
Gene Name
peptidase inhibitor 16
Gene Aliases
CD364, CRISP9, MSMBBP, PSPBP
Chromosome Location
Chr. 6: 36948263 - 36964837 on Build GRCh38
UniGene ID
Hs.25391
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199159.1 NP_001186088.1 4 693
NM_153370.2 NP_699201.2 3 798
XM_005248917.2 XP_005248974.1 3 627
XM_011514375.2 XP_011512677.1 3 627
XM_017010430.1 XP_016865919.1 3 627
GenBank mRNA AK075470.1 - 3 493
AK124589.1 - 6 2317
AK302193.1 - 3 489
AK303928.1 - 447
AK312785.1 - 3 491
AY358422.1 - 3 484
BC022399.1 - 3 492 4691115
BC035634.1 - 3 798 5217121

View Full Product Details