Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153813.2 | NP_722520.2 | 3 | 249 | |
XM_011522912.2 | XP_011521214.1 | 3 | 4098 | ||
XM_011522914.2 | XP_011521216.1 | 3 | 333 | ||
GenBank mRNA | AF488691.1 | - | 3 | 538 | |
BE219988.1 | - | 3 | 249 |