Gene Transcripts

Gene Symbol
WFDC13
Entrez Gene ID
164237
Gene Name
WAP four-disulfide core domain 13
Gene Aliases
C20orf138, WAP13, dJ601O1.3
Chromosome Location
Chr. 20: 45702016 - 45708817 on Build GRCh38
UniGene ID
Hs.406733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172005.1 NP_742002.1 4 527
GenBank mRNA AF454505.1 - 4 527

View Full Product Details