Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003327.3 | NP_003318.1 | 3 | 402 | |
XM_011542074.2 | XP_011540376.1 | 1 | 756 | ||
XM_011542075.2 | XP_011540377.1 | 3 | 869 | ||
XM_011542076.2 | XP_011540378.1 | 1 | 756 | ||
XM_017002231.1 | XP_016857720.1 | 4 | 1747 | ||
XM_017002232.1 | XP_016857721.1 | 4 | 1747 | ||
GenBank mRNA | BC105070.1 | - | 3 | 402 | |
BC105072.1 | - | 3 | 402 | 8144075 | |
S76792.1 | - | 3 | 352 |