Gene Transcripts

Gene Symbol
NTN5
Entrez Gene ID
126147
Gene Name
netrin 5
Gene Aliases
Chromosome Location
Chr. 19: 48661407 - 48673119 on Build GRCh38
UniGene ID
Hs.326217
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145807.2 NP_665806.1 4, 5 1147
XM_011526443.2 XP_011524745.1 4, 5 1185
XM_017026274.1 XP_016881763.1 3, 4 1373
GenBank mRNA BC018654.2 - 4, 5 1073
BC018697.2 - 4, 5 1073 2961115
BC021210.2 - 4, 5 1093 4137205
BC033207.1 - 4, 5 1073 4903484
BE270224.1 - 3, 4 496

View Full Product Details