Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007792.1 | NP_001007793.1 | 11 | 1314 | |
NM_001012331.1 | NP_001012331.1 | 10 | 1380 | ||
NM_002529.3 | NP_002520.2 | 11 | 1398 | ||
GenBank mRNA | AK126428.1 | - | 10 | 1252 | |
AK290759.1 | - | 10 | 1365 | ||
AK312704.1 | - | 11 | 1383 | ||
BC062580.1 | - | 11 | 1341 | ||
BC136554.1 | - | 10 | 1424 | 9020544 | |
BC144239.1 | - | 10 | 1424 | 9052759 | |
CR936794.1 | - | 12 | 1382 | ||
KJ534902.1 | - | 10 | 1324 | ||
KJ534903.1 | - | 7 | 778 | ||
M23102.1 | - | 10 | 1444 |