Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297576.1 | NP_001284505.1 | 6 | 876 | |
NM_001297577.1 | NP_001284506.1 | 5 | 674 | ||
NM_001297578.1 | NP_001284507.1 | 4 | 503 | ||
NM_003768.4 | NP_003759.1 | 4 | 569 | ||
NR_123724.1 | 3 | 395 | |||
XM_006711599.2 | XP_006711662.1 | 4 | 473 | ||
GenBank mRNA | AK095879.1 | - | 6 | 781 | |
AK311762.1 | - | 4 | 498 | ||
BC002426.2 | - | 4 | 445 | 3346270 | |
BC022554.1 | - | 4 | 556 | 4820225 | |
BT007252.1 | - | 3 | 363 | ||
CR541733.1 | - | 3 | 363 | ||
DA157938.1 | - | 4 | 439 | ||
DA532423.1 | - | 4 | 553 | ||
DA538845.1 | - | 3 | 333 | ||
Y13736.1 | - | 4 | 432 |